2-Amino-6-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile ≥95%

Supplier: Matrix Scientific

043459-500MG 043459-1G 043459-5G
101860-906EA 1109.23 USD
101860-906 101860-908 101860-910
2-Amino-6-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile ≥95%
2-Amino-6-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile

Hazard: Irritant

Formula: C₁₄H₁₀ClF₃N₄S
MW: 358.77 g/mol
Melting Pt: 199…202 °C
MDL Number: MFCD00180796
CAS Number: 321430-41-5

Order Now

Specification Test Results

Melting point 199-202°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR