O-[(2,5-Dichlorophenyl)methyl]hydroxylamine hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
2-[(Ammoniooxy)methyl]-1,4-dichlorobenzene chloride
043457-500MG
043457-1G
043457-5G
101860-894EA
747.69
USD
101860-894
101860-896
101860-898
O-[(2,5-Dichlorophenyl)methyl]hydroxylamine hydrochloride ≥95%
O-[(2,5-Dichlorophenyl)methyl]hydroxylamine hydrochloride
Hazard: Irritant
Formula:
C₇H₇Cl₂NO·HCl MW: 228.504 g/mol Melting Pt: 188…195 °C |
MDL Number:
MFCD01114576 CAS Number: 317821-71-9 |
Specification Test Results
Melting point | 188-195°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...