O-[(2,3-Dichlorophenyl)methyl]hydroxylamine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-[(Ammoniooxy)methyl]-2,3-dichlorobenzene chloride

043456-500MG 043456-1G 043456-5G
101860-888EA 747.69 USD
101860-888 101860-890 101860-892
O-[(2,3-Dichlorophenyl)methyl]hydroxylamine hydrochloride ≥95%
1-[(Ammoniooxy)methyl]-2,3-dichlorobenzene chloride

Hazard: Irritant

Formula: C₇H₇Cl₂NO·HCl
MW: 228.504 g/mol
Melting Pt: 187…192 °C
MDL Number: MFCD01114575
CAS Number: 317821-70-8

Order Now

Specification Test Results

Melting point 187-192°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR