1-[(Ammoniooxy)methyl]-2-bromobenzene chloride ≥95%
Supplier: Matrix Scientific
043454-500MG
043454-1G
043454-5G
101860-876EA
538.46
USD
101860-876
101860-878
101860-880
1-[(Ammoniooxy)methyl]-2-bromobenzene chloride ≥95%
1-[(Ammoniooxy)methyl]-2-bromobenzene chloride
Hazard: Irritant
Formula:
C₇H₉BrClNO Melting Pt: 144-146°C |
MDL Number:
MFCD01114573 CAS Number: 51572-91-9 |
Specification Test Results
Melting point | 144-146°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...