Methyl-2-amino-6-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4,5,6,7-tetrahydrothieno[2,3-c]pyridine ≥95%
Supplier: Matrix Scientific
043453-500MG
043453-1G
043453-5G
101860-870EA
646.15
USD
101860-870
101860-872
101860-874
Methyl-2-amino-6-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4,5,6,7-tetrahydrothieno[2,3-c]pyridine ≥95%
Methyl-2-amino-6-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4,5,6,7-tetrahydrothieno[2,3-c]pyridine
Hazard: Irritant
Formula:
C₁₅H₁₃ClF₃N₃O₂S MW: 391.8 g/mol Melting Pt: 149…151 °C |
MDL Number:
MFCD00180791 CAS Number: 321430-29-9 |
Specification Test Results
Melting point | 149-151°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...