4-(1H-Imidazol-1-yl)-3-nitrobenzenecarboxylic acid ≥95%
Supplier: Matrix Scientific
043452-500MG
043452-1G
043452-5G
101860-864EA
98.46
USD
101860-864
101860-866
101860-868
4-(1H-Imidazol-1-yl)-3-nitrobenzenecarboxylic acid ≥95%
4-(1H-Imidazol-1-yl)-3-nitrobenzenecarboxylic acid
Hazard: Irritant
Formula:
C₁₀H₇N₃O₄ Melting Pt: 267…269 °C |
MDL Number:
MFCD01935984 CAS Number: 167626-67-7 |
Specification Test Results
Melting point | 267-269°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...