Methyl-3-nitro-4-(1H-1,2,4-triazol-1-yl)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

043451-500MG 043451-1G 043451-5G
101860-858EA 278.46 USD
101860-858 101860-860 101860-862
Methyl-3-nitro-4-(1H-1,2,4-triazol-1-yl)benzenecarboxylate ≥95%
Methyl-3-nitro-4-(1H-1,2,4-triazol-1-yl)benzenecarboxylate

Hazard: Irritant

Formula: C₁₀H₈N₄O₄
Melting Pt: 102…104 °C
MDL Number: MFCD00141981
CAS Number: 167626-49-5

Order Now

Specification Test Results

Melting point 102-104°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR