2-(4-Pyridinyl)-4-thiazolecarboxylic acid ≥95%
Supplier: Matrix Scientific
043448-500MG
043448-1G
043448-5G
101860-846EA
24.62
USD
101860-846
101860-848
101860-850
2-(4-Pyridinyl)-4-thiazolecarboxylic acid ≥95%
2-(4-Pyridinyl)-4-thiazolecarboxylic acid
Hazard: Irritant
Formula:
C₉H₆N₂O₂S MW: 206.22 Melting Pt: 316-318°C |
MDL Number:
MFCD00171745 CAS Number: 21278-86-4 |
Specification Test Results
Melting point | 316-318°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...