N-{2-[(2-Hydroxyethyl)amino]-2-oxoethyl}benzenecarboxamide ≥95%
Supplier: Matrix Scientific
043443-500MG
043443-1G
043443-5G
101860-816EA
375.38
USD
101860-816
101860-818
101860-820
N-{2-[(2-Hydroxyethyl)amino]-2-oxoethyl}benzenecarboxamide ≥95%
N-{2-[(2-Hydroxyethyl)amino]-2-oxoethyl}benzenecarboxamide
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O₃ Melting Pt: 126…128 °C |
MDL Number:
MFCD00140890 CAS Number: 72085-01-9 |
Specification Test Results
Melting point | 126-128°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...