N-[2-(4-Aminoanilino)-2-oxoethyl]benzenecarboxamide ≥95%

Supplier: Matrix Scientific

043442-500MG 043442-1G 043442-5G
101860-810EA 375.38 USD
101860-810 101860-812 101860-814
N-[2-(4-Aminoanilino)-2-oxoethyl]benzenecarboxamide ≥95%
N-[2-(4-Aminoanilino)-2-oxoethyl]benzenecarboxamide

Hazard: Irritant

Formula: C₁₅H₁₅N₃O₂
MW: 269.31 g/mol
Melting Pt: 228…230 °C
MDL Number: MFCD00172388
CAS Number: 108717-59-5

Order Now

Specification Test Results

Melting point 228-230°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR