5,6-Dihydrobenzo[h]quinazolin-2-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 5H,6H-Benzo[h]quinazolin-2-amine

043439-500MG 043439-1G 043439-5G
101860-798EA 307.69 USD
101860-798 101860-800 101860-802
5,6-Dihydrobenzo[h]quinazolin-2-amine ≥95%
5,6-Dihydrobenzo[h]quinazolin-2-amine

Hazard: Irritant

Formula: C₁₂H₁₁N₃
MW: 197.24 g/mol
Melting Pt: 193…196 °C
MDL Number: MFCD00139519
CAS Number: 66521-84-4

Order Now

Specification Test Results

Melting point 193-196°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR