1-Allyl-2-oxo-1,2-dihydro-3-pyridinecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Oxo-1-(prop-2-en-1-yl)-1,2-dihydropyridine-3-carboxylic acid

043437-500MG 043437-1G 043437-5G
101860-788EA 220 USD
101860-788 101860-790 101860-792
1-Allyl-2-oxo-1,2-dihydro-3-pyridinecarboxylic acid ≥95%
1-Allyl-2-oxo-1,2-dihydro-3-pyridinecarboxylic acid

Hazard: Irritant

Formula: C₉H₉NO₃
MW: 179.18 g/mol
Melting Pt: 126…127 °C
MDL Number: MFCD00140660
CAS Number: 66158-33-6

Order Now

Specification Test Results

Melting point 126-127°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR