2-(6-Chloro-3-pyridazinyl)-2-(2,6-dichlorophenyl)acetonitrile ≥95%

Supplier: Matrix Scientific

043435-500MG 043435-1G 043435-5G
101860-776EA 181.54 USD
101860-776 101860-778 101860-780
2-(6-Chloro-3-pyridazinyl)-2-(2,6-dichlorophenyl)acetonitrile ≥95%
2-(6-Chloro-3-pyridazinyl)-2-(2,6-dichlorophenyl)acetonitrile

Hazard: Irritant

Formula: C₁₂H₆Cl₃N₃
Melting Pt: 142-144°C
MDL Number: MFCD00202119
CAS Number: 209412-06-6

Order Now

Specification Test Results

Melting point 142-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR