3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenol ≥95%
Supplier: Matrix Scientific
043434-500MG
043434-1G
043434-5G
101860-770EA
375.38
USD
101860-770
101860-772
101860-774
3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenol ≥95%
3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenol
Hazard: Irritant
Formula:
C₁₂H₇ClF₃NO₂ MW: 289.64 g/mol Melting Pt: 100…102 °C |
MDL Number:
MFCD00172750 CAS Number: 95711-07-2 |
Specification Test Results
Melting point | 100-102°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...