Methyl-4-chloro-6-oxo-1-phenyl-1,6-dihydro-3-pyridazinecarboxylate ≥95%

Supplier: Matrix Scientific

043433-500MG 043433-1G 043433-5G
101860-764EA 461.54 USD
101860-764 101860-766 101860-768
Methyl-4-chloro-6-oxo-1-phenyl-1,6-dihydro-3-pyridazinecarboxylate ≥95%
Methyl-4-chloro-6-oxo-1-phenyl-1,6-dihydro-3-pyridazinecarboxylate

Hazard: Irritant

Formula: C₁₂H₉ClN₂O₃
MW: 264.67 g/mol
Melting Pt: 104…106 °C
MDL Number: MFCD00141141
CAS Number: 129109-17-7

Order Now

Specification Test Results

Melting point 104-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR