4-(2,4-Dichlorobenzoyl)-1-methyl-1H-pyrrole-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
043432-500MG
043432-1G
043432-5G
101860-758EA
747.69
USD
101860-758
101860-760
101860-762
4-(2,4-Dichlorobenzoyl)-1-methyl-1H-pyrrole-2-carboxylic acid ≥95%
4-(2,4-Dichlorobenzoyl)-1-methyl-1H-pyrrole-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₉Cl₂NO₃ Melting Pt: 206…208 °C |
MDL Number:
MFCD00173071 CAS Number: 320419-40-7 |
Specification Test Results
Melting point | 206-208°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...