3-(1-Nitroethyl)-2-benzofuran-1(3H)-one ≥95%
Supplier: Matrix Scientific
043431-500MG
043431-1G
043431-5G
101860-752EA
273.85
USD
101860-752
101860-754
101860-756
3-(1-Nitroethyl)-2-benzofuran-1(3H)-one ≥95%
3-(1-Nitroethyl)-2-benzofuran-1(3H)-one
Hazard: Irritant
Formula:
C₁₀H₉NO₄ Melting Pt: 99…102 °C |
MDL Number:
MFCD03001201 CAS Number: 79017-08-6 |
Specification Test Results
Melting point | 99-102°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...