2-(6-Chloro-3-pyridazinyl)-2-(2-fluorophenyl)acetonitrile ≥95%

Supplier: Matrix Scientific

043429-500MG 043429-1G 043429-5G
101860-740EA 181.54 USD
101860-740 101860-742 101860-744
2-(6-Chloro-3-pyridazinyl)-2-(2-fluorophenyl)acetonitrile ≥95%
2-(6-Chloro-3-pyridazinyl)-2-(2-fluorophenyl)acetonitrile

Hazard: Irritant

Formula: C₁₂H₇ClFN₃
Melting Pt: 69-71°C
MDL Number: MFCD00202113
CAS Number: 154419-42-8

Order Now

Specification Test Results

Melting point 69-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR