5-(Carboxymethyl)-1-(2,4-dichlorophenyl)-1H-1,2,3-triazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

043427-500MG 043427-1G 043427-5G
101860-730EA 747.69 USD
101860-730 101860-732 101860-734
5-(Carboxymethyl)-1-(2,4-dichlorophenyl)-1H-1,2,3-triazole-4-carboxylic acid ≥95%
5-(Carboxymethyl)-1-(2,4-dichlorophenyl)-1H-1,2,3-triazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₇Cl₂N₃O₄
Melting Pt: 122…124 °C
MDL Number: MFCD00127287
CAS Number: 320418-32-4

Order Now

Specification Test Results

Melting point 122-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR