Methyl-4-(4-fluorophenoxy)-3-nitrobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-(4-fluorophenoxy)-3-nitrobenzoate

043425-500MG 043425-1G 043425-5G
101860-724EA 375.38 USD
101860-724 101860-726 101860-728
Methyl-4-(4-fluorophenoxy)-3-nitrobenzenecarboxylate ≥95%
Methyl-4-(4-fluorophenoxy)-3-nitrobenzenecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₀FNO₅
MW: 291.24 g/mol
Melting Pt: 101…103 °C
MDL Number: MFCD00138765
CAS Number: 320417-11-6

Order Now

Specification Test Results

Melting point 101-103°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR