3-(Dimethylamino)-1-phenyl-2-propen-1-one ≥95%

Supplier: Matrix Scientific

043423-1G 043423-5G
101860-714EA 58.46 USD
101860-714 101860-716
3-(Dimethylamino)-1-phenyl-2-propen-1-one ≥95%
3-(Dimethylamino)-1-phenyl-2-propen-1-one

Hazard: Irritant

Formula: C₁₁H₁₃NO
Melting Pt: 91…93 °C
MDL Number: MFCD00121193
CAS Number: 1201-93-0

Order Now

Specification Test Results

Melting point 91-93°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR