4-(4-Methylphenoxy)-3-nitrobenzaldehyde ≥95%
Supplier: Matrix Scientific
043422-500MG
043422-1G
043422-5G
101860-706EA
181.54
USD
101860-706
101860-708
101860-710
4-(4-Methylphenoxy)-3-nitrobenzaldehyde ≥95%
4-(4-Methylphenoxy)-3-nitrobenzaldehyde
Hazard: Irritant
Formula:
C₁₄H₁₁NO₄ Melting Pt: 108-109°C |
MDL Number:
MFCD06659537 CAS Number: 750641-06-6 |
Specification Test Results
Melting point | 108-109°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...