4-(3-Chlorophenoxy)-3-nitrobenzenecarbaldehyde ≥95%
Supplier: Matrix Scientific
043421-500MG
043421-1G
043421-5G
101860-700EA
375.38
USD
101860-700
101860-702
101860-704
4-(3-Chlorophenoxy)-3-nitrobenzenecarbaldehyde ≥95%
4-(3-Chlorophenoxy)-3-nitrobenzenecarbaldehyde
Hazard: Irritant
Formula:
C₁₃H₈ClNO₄ Melting Pt: 80…82 °C |
MDL Number:
MFCD00138762 CAS Number: 320417-03-6 |
Specification Test Results
Melting point | 80-82°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...