[1,2,4]Triazolo[1,5-a]pyrimidin-7-amine ≥95%

Supplier: Matrix Scientific

043420-500MG 043420-1G 043420-5G
101860-694EA 150.77 USD
101860-694 101860-696 101860-698
[1,2,4]Triazolo[1,5-a]pyrimidin-7-amine ≥95%
[1,2,4]Triazolo[1,5-a]pyrimidin-7-amine

Hazard: Irritant

Formula: C₅H₅N₅
Melting Pt: 278…280 °C
MDL Number: MFCD00232059
CAS Number: 5915-16-2

Order Now

Specification Test Results

Melting point 278-280°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR