3-Nitro-4-[3-(trifluoromethyl)phenoxy]benzenecarbaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Nitro-4-[3-(trifluoromethyl)phenoxy]benzaldehyde
043418-500MG
043418-1G
043418-5G
101860-682EA
375.38
USD
101860-682
101860-684
101860-686
3-Nitro-4-[3-(trifluoromethyl)phenoxy]benzenecarbaldehyde ≥95%
3-Nitro-4-[3-(trifluoromethyl)phenoxy]benzenecarbaldehyde
Hazard: Irritant
Formula:
C₁₄H₈F₃NO₄ MW: 311.22 g/mol Melting Pt: 107…108 °C |
MDL Number:
MFCD00138749 CAS Number: 320416-66-8 |
Specification Test Results
Melting point | 107-108°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...