4-(4-Fluorophenoxy)-3-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

043417-500MG 043417-1G 043417-5G
101860-676EA 375.38 USD
101860-676 101860-678 101860-680
4-(4-Fluorophenoxy)-3-nitrobenzaldehyde ≥95%
4-(4-Fluorophenoxy)-3-nitrobenzaldehyde

Hazard: Irritant

Formula: C₁₃H₈FNO₄
MDL Number: MFCD00138746
CAS Number: 320416-60-2

Order Now

Specification Test Results

Melting point 89-91°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR