Methyl-4-(4-chlorophenoxy)-3-nitrobenzenecarboxylate ≥95%
Supplier: Matrix Scientific
043416-500MG
043416-1G
043416-5G
101860-670EA
375.38
USD
101860-670
101860-672
101860-674
Methyl-4-(4-chlorophenoxy)-3-nitrobenzenecarboxylate ≥95%
Methyl-4-(4-chlorophenoxy)-3-nitrobenzenecarboxylate
Hazard: Irritant
Formula:
C₁₄H₁₀ClNO₅ Melting Pt: 125…127 °C |
MDL Number:
MFCD00138745 CAS Number: 320416-58-8 |
Specification Test Results
Melting point | 125-127°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...