4-(4-Chlorophenoxy)-3-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

043415-500MG 043415-1G 043415-5G
101860-664EA 375.38 USD
101860-664 101860-666 101860-668
4-(4-Chlorophenoxy)-3-nitrobenzaldehyde ≥95%
4-(4-Chlorophenoxy)-3-nitrobenzaldehyde

Hazard: Irritant

Formula: C₁₃H₈ClNO₄
Melting Pt: 117-119°C
MDL Number: MFCD01113126
CAS Number: 320416-35-1

Order Now

Specification Test Results

Melting point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR