2-(6-Chloro-2-pyridinyl)-1-hydrazinecarboxamide ≥95%

Supplier: Matrix Scientific

043411-500MG 043411-1G 043411-5G
101860-640EA 375.38 USD
101860-640 101860-642 101860-644
2-(6-Chloro-2-pyridinyl)-1-hydrazinecarboxamide ≥95%
2-(6-Chloro-2-pyridinyl)-1-hydrazinecarboxamide

Hazard: Irritant

Formula: C₆H₇ClN₄O
Melting Pt: 226…228 °C
MDL Number: MFCD00138719
CAS Number: 320415-91-6

Order Now

Specification Test Results

Melting point 226-228°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR