2-Chloroquinoxaline ≥95%

Supplier: Matrix Scientific

043410-500MG 043410-1G 043410-5G
101860-634EA 15.38 USD
101860-634 101860-636 101860-638
2-Chloroquinoxaline ≥95%
2-Chloroquinoxaline

Hazard: Irritant

Formula: C₈H₅ClN₂
Melting Pt: 46 °C
MDL Number: MFCD00043907
CAS Number: 1448-87-9

Order Now

Specification Test Results

Melting point 46°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR