Methyl-3-(2-chlorophenyl)-5-methyl-4-isoxazolecarboxylate ≥95%
Supplier: Matrix Scientific
043407-500MG
043407-1G
043407-5G
101860-622EA
304.62
USD
101860-622
101860-624
101860-626
Methyl-3-(2-chlorophenyl)-5-methyl-4-isoxazolecarboxylate ≥95%
Methyl-3-(2-chlorophenyl)-5-methyl-4-isoxazolecarboxylate
Hazard: Irritant
Formula:
C₁₂H₁₀ClNO₃ MW: 251.67 g/mol Melting Pt: 56…58 °C |
MDL Number:
MFCD00214711 CAS Number: 4357-94-2 |
Specification Test Results
Melting point | 56-58°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...