3-(Nitromethyl)-2-benzofuran-1(3H)-one ≥95%

Supplier: Matrix Scientific

043405-500MG 043405-1G 043405-5G
101860-616EA 184.62 USD
101860-616 101860-618 101860-620
3-(Nitromethyl)-2-benzofuran-1(3H)-one ≥95%
3-(Nitromethyl)-2-benzofuran-1(3H)-one

Hazard: Irritant

Formula: C₉H₇NO₄
Melting Pt: 126…129 °C
MDL Number: MFCD00140496
CAS Number: 3598-68-3

Order Now

Specification Test Results

Melting point 126-129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR