2-Hydrazino-5-nitro-1H-1,3-benzimidazole ≥95%

Supplier: Matrix Scientific

043402-500MG 043402-1G 043402-5G
101860-602EA 278.46 USD
101860-602 101860-604 101860-606
2-Hydrazino-5-nitro-1H-1,3-benzimidazole ≥95%
2-Hydrazino-5-nitro-1H-1,3-benzimidazole

Hazard: Irritant

Formula: C₇H₇N₅O₂
Melting Pt: 235…236 °C
MDL Number: MFCD08276961
CAS Number: 91467-48-0

Order Now

Specification Test Results

Melting point 235-236°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR