1-[2-Amino-5-(1-pyrrolidinyl)phenyl]-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 2'-Amino-5'-(pyrrolidin-1-yl)acetophenone

043400-500MG 043400-1G 043400-5G
101860-596EA 556.92 USD
101860-596 101860-598 101860-600
1-[2-Amino-5-(1-pyrrolidinyl)phenyl]-1-ethanone ≥95%
1-[2-Amino-5-(1-pyrrolidinyl)phenyl]-1-ethanone

Hazard: Irritant

Formula: C₁₂H₁₆N₂O
MW: 204.27 g/mol
Melting Pt: 79…81 °C
MDL Number: MFCD08443939
CAS Number: 56915-84-5

Order Now

Specification Test Results

Melting point 79-81°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR