1-[2-Amino-5-(3-methylpiperidino)phenyl]-1-ethanone ≥95%
Supplier: Matrix Scientific
Synonyms:
2'-Amino-5'-(3-methylpiperidin-1-yl)acetophenone
043398-500MG
043398-1G
043398-5G
101860-584EA
360
USD
101860-584
101860-586
101860-588
1-[2-Amino-5-(3-methylpiperidino)phenyl]-1-ethanone ≥95%
1-[2-Amino-5-(3-methylpiperidino)phenyl]-1-ethanone
Hazard: Irritant
Formula:
C₁₄H₂₀N₂O MW: 232.33 g/mol Melting Pt: 83…85 °C |
MDL Number:
MFCD08276954 CAS Number: 886361-37-1 |
Specification Test Results
Melting point | 83-85°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...