4-(1,3-Oxazol-5-yl)aniline ≥95%
Supplier: Matrix Scientific
043397-500MG
043397-1G
043397-5G
101860-578EA
278.46
USD
101860-578
101860-580
101860-582
4-(1,3-Oxazol-5-yl)aniline ≥95%
4-(1,3-Oxazol-5-yl)aniline
Hazard: Irritant
Formula:
C₉H₈N₂O MW: 160.1700 g/mol Melting Pt: 143-145°C |
MDL Number:
MFCD00052192 CAS Number: 1008-95-3 |
Specification Test Results
Melting point | 143-145°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...