1-[2-Amino-5-(4-hydroxypiperidino)phenyl]-1-ethanone ≥95%
Supplier: Matrix Scientific
043396-500MG
043396-1G
043396-5G
101860-572EA
921.54
USD
101860-572
101860-574
101860-576
1-[2-Amino-5-(4-hydroxypiperidino)phenyl]-1-ethanone ≥95%
1-[2-Amino-5-(4-hydroxypiperidino)phenyl]-1-ethanone
Hazard: Irritant
Formula:
C₁₃H₁₈N₂O₂ MW: 234.3 g/mol Melting Pt: 184…186 °C |
MDL Number:
MFCD08276932 CAS Number: 359841-43-3 |
Specification Test Results
Melting point | 184-186°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...