5-Bromo-2-piperidinobenzenecarbaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-2-piperidin-1-ylbenzaldehyde
043393-500MG
043393-1G
043393-5G
101860-554EA
278.46
USD
101860-554
101860-556
101860-558
5-Bromo-2-piperidinobenzenecarbaldehyde ≥95%
5-Bromo-2-piperidinobenzenecarbaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₄BrNO MW: 268.16 g/mol Melting Pt: 72…75 °C |
MDL Number:
MFCD08276957 CAS Number: 742099-33-8 |
Specification Test Results
Melting point | 72-75°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...