4-(1,4-Thiazinan-4-yl)benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Thiomorpholin-4-yl)benzaldehyde

043391-500MG 043391-1G
101860-544EA 200 USD
101860-544 101860-546
4-(1,4-Thiazinan-4-yl)benzenecarbaldehyde ≥95%
4-(1,4-Thiazinan-4-yl)benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₁₃NOS
MW: 207.3 g/mol
Melting Pt: 63…65 °C
MDL Number: MFCD08276967
CAS Number: 27913-94-6

Order Now

Specification Test Results

Melting point 63-65°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR