Ethyl-5-(trifluoromethyl)picolinate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-(trifluoromethyl)-2-pyridinecarboxylate

043386-1G 043386-5G
101860-530EA 80 USD
101860-530 101860-532
Ethyl-5-(trifluoromethyl)picolinate ≥95%
Ethyl 5-(trifluoromethyl)picolinate

Hazard: Irritant

Formula: C₉H₈F₃NO₂
MW: 219.16 g/mol
Melting Pt: 45-48°C
MDL Number: MFCD11100197
CAS Number: 128072-94-6

Order Now

Specification Test Results

Melting point 45-48°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR