2-(Chloromethyl)-5-(4-nitrophenyl)-1,3,4-oxadiazole ≥95%
Supplier: Matrix Scientific
043384-500MG
043384-1G
043384-5G
101860-516EA
556.92
USD
101860-516
101860-518
101860-520
2-(Chloromethyl)-5-(4-nitrophenyl)-1,3,4-oxadiazole ≥95%
2-(Chloromethyl)-5-(4-nitrophenyl)-1,3,4-oxadiazole
Hazard: Irritant
Formula:
C₉H₆ClN₃O₃ Melting Pt: 133…135 °C |
MDL Number:
MFCD00775433 CAS Number: 50677-30-0 |
Specification Test Results
Melting point | 133-135°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...