6-[5-(Chloromethyl)-1,3,4-oxadiazol-2-yl]-2,3-dimethylquinoxaline ≥95%

Supplier: Matrix Scientific

043383-500MG 043383-1G
101860-512EA 921.54 USD
101860-512 101860-514
6-[5-(Chloromethyl)-1,3,4-oxadiazol-2-yl]-2,3-dimethylquinoxaline ≥95%
6-[5-(Chloromethyl)-1,3,4-oxadiazol-2-yl]-2,3-dimethylquinoxaline

Hazard: Irritant

Formula: C₁₃H₁₁ClN₄O
Melting Pt: 183…185 °C
MDL Number: MFCD07366536
CAS Number: 885949-52-0

Order Now

Specification Test Results

Melting point 183-185°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR