2-(Chloromethyl)-5-(3,4-dichlorophenyl)-1,3,4-oxadiazole ≥95%
Supplier: Matrix Scientific
043381-500MG
043381-1G
043381-5G
101860-506EA
747.69
USD
101860-506
101860-508
101860-510
2-(Chloromethyl)-5-(3,4-dichlorophenyl)-1,3,4-oxadiazole ≥95%
2-(Chloromethyl)-5-(3,4-dichlorophenyl)-1,3,4-oxadiazole
Hazard: Irritant
Formula:
C₉H₅Cl₃N₂O Melting Pt: 120…122 °C |
MDL Number:
MFCD01886041 CAS Number: 33575-81-4 |
Specification Test Results
Melting point | 120-122°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...