Ethyl-4-morpholinobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-(morpholin-4-yl)benzoate, Ethyl 4-Morpholinobenzoate

043375-500MG 043375-1G 043375-5G
101860-472EA 150.77 USD
101860-472 101860-474 101860-476
Ethyl-4-morpholinobenzenecarboxylate ≥95%
Ethyl-4-morpholinobenzenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₇NO₃
MW: 235.29 g/mol
Melting Pt: 86…88 °C
MDL Number: MFCD06204958
CAS Number: 19614-15-4

Order Now

Specification Test Results

Melting point 86-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR