2-Amino-5-(2-chlorophenyl)-4-thiazolecarboxylic acid ≥95%

Supplier: Matrix Scientific

043364-500MG 043364-1G 043364-5G
101860-444EA 278.46 USD
101860-444 101860-446 101860-448
2-Amino-5-(2-chlorophenyl)-4-thiazolecarboxylic acid ≥95%
2-Amino-5-(2-chlorophenyl)-4-thiazolecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₇ClN₂O₂S
Melting Pt: 246°C
MDL Number: MFCD06200928
CAS Number: 886361-44-0

Order Now

Specification Test Results

Melting point 246°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR