2-Chloro-N-(tetrahydro-2-furanylmethyl)acetamide ≥95%
Supplier: Matrix Scientific
043361-500MG
043361-1G
043361-5G
101860-434EA
181.54
USD
101860-434
101860-436
101860-438
2-Chloro-N-(tetrahydro-2-furanylmethyl)acetamide ≥95%
2-Chloro-N-(tetrahydro-2-furanylmethyl)acetamide
Hazard: Irritant
Formula:
C₇H₁₂ClNO₂ Melting Pt: 42…45 °C |
MDL Number:
MFCD01182294 CAS Number: 39089-62-8 |
Specification Test Results
Melting point | 42-45°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...