Methyl-5-methylpyrazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-methyl-1H-pyrazole-3-carboxylate, Methyl 5-methyl[1H]pyrazole-3-carboxylate, 5-Methylpyrazole-3-carboxylic Acid Methyl Ester

043360-500MG 043360-1G 043360-5G
101860-428EA 30.77 USD
101860-428 101860-430 101860-432
Methyl-5-methylpyrazole-3-carboxylate ≥95%
Methyl-5-methylpyrazole-3-carboxylate

Hazard: Irritant

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Melting Pt: 82…84 °C
MDL Number: MFCD03778987
CAS Number: 25016-17-5

Order Now

Specification Test Results

Melting point 82-84°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR