2-Ethyl-1H-imidazole-4,5-dicarbonitrile ≥95%

Supplier: Matrix Scientific

043359-500MG 043359-1G 043359-5G
101860-422EA 278.46 USD
101860-422 101860-424 101860-426
2-Ethyl-1H-imidazole-4,5-dicarbonitrile ≥95%
2-Ethyl-1H-imidazole-4,5-dicarbonitrile

Hazard: Irritant

Formula: C₇H₆N₄
Melting Pt: 172…175 °C
MDL Number: MFCD06496123
CAS Number: 57610-38-5

Order Now

Specification Test Results

Melting point 172-175°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR