4,5-Dicyano-2-phenylimidazole ≥95%

Supplier: Matrix Scientific

043358-500MG 043358-1G 043358-5G
101860-416EA 278.46 USD
101860-416 101860-418 101860-420
4,5-Dicyano-2-phenylimidazole ≥95%
4,5-Dicyano-2-phenylimidazole

Hazard: Irritant

Formula: C₁₁H₆N₄
MW: 194.19 g/mol
Melting Pt: 192…196 °C (dec.)
MDL Number: MFCD05863325
CAS Number: 50847-06-8
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 192-196°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR