(4-Chlorophenyl)(piperidin-4-yl)methanone hydrochloride ≥95%

Supplier: Matrix Scientific

043356-500MG 043356-1G 043356-5G
101860-410EA 921.54 USD
101860-410 101860-412 101860-414
(4-Chlorophenyl)(piperidin-4-yl)methanone hydrochloride ≥95%
(4-Chlorophenyl)(piperidin-4-yl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅Cl₂NO
MW: 260.1600 g/mol
Melting Pt: 236…237 °C
MDL Number: MFCD00053028
CAS Number: 55695-51-7

Order Now

Specification Test Results

Melting point 236-237°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR